COMH42122702 Combi-Blocks, Inc.

(2R)-3-(3-cyanophenyl)-2-acetamidopropanoic acid

  • MDL number MFCD30475860
  • Cas Number : 367453-00-7
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9740
Availability details for
Quantity
  • Availability
  • Add to Cart