COMH42122705 Combi-Blocks, Inc.

(Z)-N'-hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzene-1-carboximidamide

  • MDL number MFCD00830779
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9795
Availability details for
Quantity
  • Availability
  • Add to Cart