COMH4212270B Combi-Blocks, Inc.

4-[2-(4-bromophenoxy)ethyl]pyridine

  • MDL number MFCD12799513
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1132
Availability details for
Quantity
  • Availability
  • Add to Cart