COMH4212270C Combi-Blocks, Inc.

N-methoxy-N,2-dimethylpyridine-3-carboxamide

  • MDL number MFCD15526807
  • Cas Number : 469864-28-6
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1146

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1146
Availability details for
Quantity
  • Availability
  • Add to Cart