COMH42122711 Combi-Blocks, Inc.

2-(4-methylpiperazin-1-yl)pyridine-4-carboxylic acid

  • MDL number MFCD07801144
  • Cas Number : 876717-56-5
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1460

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1460
Availability details for
Quantity
  • Availability
  • Add to Cart