COMH42122713 Combi-Blocks, Inc.

6-Methyl-2H-[1,2,4]triazolo[4,3-b]pyridazin-3-one

  • MDL number MFCD00197444
  • Cas Number : 39030-53-0
  • Molecular Weight : 150.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1842

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1842
Availability details for
Quantity
  • Availability
  • Add to Cart