COMH42122715 Combi-Blocks, Inc.

tert-Butyl N-(6-fluoropyridin-2-yl)carbamate

  • MDL number MFCD11976435
  • Cas Number : 891856-23-8
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2006

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2006
Availability details for
Quantity
  • Availability
  • Add to Cart