COMH42122716 Combi-Blocks, Inc.

N-Cyclopropyl-2-methyl-5-nitrobenzenesulfonamide

  • MDL number MFCD09583904
  • Cas Number : 931882-97-2
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4955
Availability details for
Quantity
  • Availability
  • Add to Cart