COMH4212271D Combi-Blocks, Inc.

tert-Butyl 2-[(4-methoxyphenyl)carbonyl]piperidine-1-carboxylate

  • MDL number MFCD24535902
  • Cas Number : 1824432-70-3
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-4710

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-4710
Availability details for
Quantity
  • Availability
  • Add to Cart