COMH42122723 Combi-Blocks, Inc.

3-(2,2-Dimethylpropoxy)benzoic acid

  • MDL number MFCD06801151
  • Cas Number : 883544-24-9
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5388
Availability details for
Quantity
  • Availability
  • Add to Cart