COMH42122724 Combi-Blocks, Inc.

2-(2,2-Dimethylpropoxy)benzaldehyde

  • MDL number MFCD32065790
  • Cas Number : 1688705-80-7
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5389

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5389
Availability details for
Quantity
  • Availability
  • Add to Cart