COMH42122725 Combi-Blocks, Inc.

2-(2,2-Dimethylpropoxy)benzoic acid

  • MDL number MFCD32065791
  • Cas Number : 2163563-77-5
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5390
Availability details for
Quantity
  • Availability
  • Add to Cart