COMH4212275A Combi-Blocks, Inc.

Methyl 1-methylazetidine-3-carboxylate

  • MDL number MFCD32065792
  • Cas Number : 2115351-22-7
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6090
Availability details for
Quantity
  • Availability
  • Add to Cart