COMH4212275D Combi-Blocks, Inc.

Methyl 3-(3-hydroxypropoxy)propanoate

  • MDL number MFCD28969631
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6817

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6817
Availability details for
Quantity
  • Availability
  • Add to Cart