COMH4212275E Combi-Blocks, Inc.

1-Benzyl-3-methylpyrrolidin-2-one

  • MDL number MFCD30290809
  • Cas Number : 108303-99-7
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6877
Availability details for
Quantity
  • Availability
  • Add to Cart