COMH42122760 Combi-Blocks, Inc.

Benzyl N-(5-bromo-2,3-dihydro-1H-inden-2-yl)carbamate

  • MDL number MFCD26395176
  • Cas Number : 892492-33-0
  • Molecular Weight : 346.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7210

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7210
Availability details for
Quantity
  • Availability
  • Add to Cart