COMH42123E7F Combi-Blocks, Inc.

[3-(2-Methylprop-2-en-1-yl)phenyl]boronic acid

  • MDL number MFCD32065800
  • Molecular Weight : 176.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7304
Availability details for
Quantity
  • Availability
  • Add to Cart