COMH42123E82 Combi-Blocks, Inc.

4-(2-Aminoethoxy)butan-2-one; trifluoroacetic acid

  • MDL number MFCD32065806
  • Cas Number : 2365418-59-1
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7358

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7358
Availability details for
Quantity
  • Availability
  • Add to Cart