COMH42123E83 Combi-Blocks, Inc.

1-Benzyl-3-methylpyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD32065808
  • Cas Number : 2365419-16-3
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7374
Availability details for
Quantity
  • Availability
  • Add to Cart