COMH42123E8B Combi-Blocks, Inc.

tert-Butyl 2-(5-methyl-1,2,4-oxadiazol-3-yl)acetate

  • MDL number MFCD32065824
  • Cas Number : 2365420-05-7
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7508
Availability details for
Quantity
  • Availability
  • Add to Cart