COMH42123E8D Combi-Blocks, Inc.

4-Bromo-2,3,5,6-tetramethylbenzaldehyde

  • MDL number MFCD32065804
  • Cas Number : 88174-44-1
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7338

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7338
Availability details for
Quantity
  • Availability
  • Add to Cart