COMH42123E8F Combi-Blocks, Inc.

N-(2-Phenylethyl)pyrrolidine-3-carboxamide

  • MDL number MFCD17271859
  • Cas Number : 1230100-87-4
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7342

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7342
Availability details for
Quantity
  • Availability
  • Add to Cart