COMH42123E92 Combi-Blocks, Inc.

N-Isopropylpyrrolidine-3-carboxamide

  • MDL number MFCD17271798
  • Cas Number : 1339569-89-9
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7345

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7345
Availability details for
Quantity
  • Availability
  • Add to Cart