COMH42123E93 Combi-Blocks, Inc.

N-Cyclopropylpyrrolidine-3-carboxamide

  • MDL number MFCD17271803
  • Cas Number : 960491-77-4
  • Molecular Weight : 154.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7346
Availability details for
Quantity
  • Availability
  • Add to Cart