COMH42123E94 Combi-Blocks, Inc.

Benzyl 4-methyl-3,6-dihydro-2H-pyridine-1-carboxylate

  • MDL number MFCD28533700
  • Cas Number : 1785555-47-6
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7350

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7350
Availability details for
Quantity
  • Availability
  • Add to Cart