COMH42123E99 Combi-Blocks, Inc.

tert-Butyl 4-(1-ethoxy-1-oxopropan-2-ylidene)piperidine-1-carboxylate

  • MDL number MFCD21607048
  • Cas Number : 473837-03-5
  • Molecular Weight : 283.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7361
Availability details for
Quantity
  • Availability
  • Add to Cart