COMH42123E9A Combi-Blocks, Inc.

2-{1-[(tert-Butoxy)carbonyl]piperidin-4-ylidene}propanoic acid

  • MDL number MFCD21607052
  • Cas Number : 1824085-55-3
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7362
Availability details for
Quantity
  • Availability
  • Add to Cart