COMH42123E9F Combi-Blocks, Inc.

2-Bromo-3,5-dichloropyridine-4-carbaldehyde

  • MDL number MFCD16606939
  • Cas Number : 1227584-22-6
  • Molecular Weight : 254.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7380
Availability details for
Quantity
  • Availability
  • Add to Cart