COMH42123EA0 Combi-Blocks, Inc.

4-Bromo-3-methoxyisobenzofuran-1(3H)-one

  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7381

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7381
Availability details for
Quantity
  • Availability
  • Add to Cart