COMH42123EA1 Combi-Blocks, Inc.

tert-Butyl N-{2-[(4-bromo-2-fluorophenyl)formamido]ethyl}carbamate

  • MDL number MFCD32065809
  • Molecular Weight : 361.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7385

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7385
Availability details for
Quantity
  • Availability
  • Add to Cart