COMH42123EA5 Combi-Blocks, Inc.

2-Chloro-3-methoxypyridine-4-carboxylic acid

  • MDL number MFCD13185828
  • Cas Number : 1214344-24-7
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7400
Availability details for
Quantity
  • Availability
  • Add to Cart