COMH42123EA6 Combi-Blocks, Inc.

benzyl N-{2-[methoxy(methyl)carbamoyl]ethyl}carbamate

  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7310
Availability details for
Quantity
  • Availability
  • Add to Cart