COMH42123EA8 Combi-Blocks, Inc.

2-(Benzyloxy)-1-(4-methylpiperazin-1-yl)ethanone

  • MDL number MFCD21366716
  • Cas Number : 1455171-49-9
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7411

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7411
Availability details for
Quantity
  • Availability
  • Add to Cart