COMH42123EA9 Combi-Blocks, Inc.

4-(Benzyloxy)-2-bromopyridine

  • MDL number MFCD15523325
  • Cas Number : 220585-13-7
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7417
Availability details for
Quantity
  • Availability
  • Add to Cart