COMH42123EAA Combi-Blocks, Inc.

2-Bromo-4-isopropoxypyridine

  • MDL number MFCD11869639
  • Cas Number : 1142194-66-8
  • Molecular Weight : 216.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7418
Availability details for
Quantity
  • Availability
  • Add to Cart