COMH42123EB1 Combi-Blocks, Inc.

3-Chloro-2,4-dimethoxypyridine

  • MDL number MFCD00235117
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7435
Availability details for
Quantity
  • Availability
  • Add to Cart