COMH42123EB2 Combi-Blocks, Inc.

2-Bromo-1-(bromomethyl)-4-tert-butylbenzene

  • MDL number MFCD09029650
  • Cas Number : 246139-76-4
  • Molecular Weight : 306.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7440

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7440
Availability details for
Quantity
  • Availability
  • Add to Cart