COMH42123EB3 Combi-Blocks, Inc.

tert-Butyl N-[(3R)-1-methylpiperidin-3-yl]carbamate

  • MDL number MFCD28402661
  • Cas Number : 1001354-56-8
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7312
Availability details for
Quantity
  • Availability
  • Add to Cart