COMH42123EB5 Combi-Blocks, Inc.

4-tert-Butyl-1-methyl-2-nitrobenzene

  • MDL number MFCD09702156
  • Cas Number : 62559-08-4
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7443

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7443
Availability details for
Quantity
  • Availability
  • Add to Cart