COMH42123EBA Combi-Blocks, Inc.

Ethyl 4-(N'-hydroxycarbamimidoyl)benzoate

  • Cas Number : 40069-10-1
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7455
Availability details for
Quantity
  • Availability
  • Add to Cart