COMH42123EC2 Combi-Blocks, Inc.

5,6-dimethyl-4,5-dihydro-2H-pyridazin-3-one

  • MDL number MFCD31561679
  • Cas Number : 811860-95-4
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7330
Availability details for
Quantity
  • Availability
  • Add to Cart