COMH42123EC6 Combi-Blocks, Inc.

5-Acetyl-2-ethoxybenzoic acid

  • MDL number MFCD1453634
  • Cas Number : 147676-95-7
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7494
Availability details for
Quantity
  • Availability
  • Add to Cart