COMH42123EC9 Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenyl)propanoic acid

  • MDL number MFCD26912288
  • Cas Number : 1565748-29-9
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7331

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7331
Availability details for
Quantity
  • Availability
  • Add to Cart