COMH42123ECA Combi-Blocks, Inc.

tert-Butyl 2-(-N'-hydroxycarbamimidoyl)acetate

  • MDL number MFCD32065823
  • Cas Number : 218278-67-2
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7507

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7507
Availability details for
Quantity
  • Availability
  • Add to Cart