COMH42123ECC Combi-Blocks, Inc.

tert-Butyl 2-(5-isopropyl-1,2,4-oxadiazol-3-yl)acetate

  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7509
Availability details for
Quantity
  • Availability
  • Add to Cart