COMH42123ED2 Combi-Blocks, Inc.

1-Boc-2-((5-bromopyridin-3-yloxy)methyl)pyrrolidine

  • MDL number MFCD19382453
  • Cas Number : 1311254-65-5
  • Molecular Weight : 357.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1941

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1941
Availability details for
Quantity
  • Availability
  • Add to Cart