COMH42123ED3 Combi-Blocks, Inc.

1-Cyclobutyl-6-methoxy-1h-indole-3-carbonitrile

  • MDL number MFCD21607442
  • Cas Number : 944563-38-6
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2182

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2182
Availability details for
Quantity
  • Availability
  • Add to Cart