COMH42123EEC Combi-Blocks, Inc.

1-Naphthylamine-3,6,8-trisulfonic acid disodium salt hydrate

  • MDL number MFCD08273764
  • Cas Number : 5398-34-5
  • Molecular Weight : 445.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7899
Availability details for
Quantity
  • Availability
  • Add to Cart