COMH42123F13 Combi-Blocks, Inc.

3-(4-Carbamoylphenoxy)propionic acid

  • MDL number MFCD09948835
  • Cas Number : 111140-92-2
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1693
Availability details for
Quantity
  • Availability
  • Add to Cart