COMH42123F21 Combi-Blocks, Inc.

2-Phenyl-2h-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD01104763
  • Cas Number : 3213-80-7
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2128
Availability details for
Quantity
  • Availability
  • Add to Cart